Learn More
Axygen™ Automation Tips for PerkinElmer™, 384-Well
Ideal for Janus™ MiniTrak, PlateTrak™ and Evolution P3. Axygen™ Automation Tips for PerkinElmer™, 384-Well are racks of non-filtered, 30μL robotic pipette tips. Constructed of clear polypropyle to stringent specifications. Available in sterile and non-sterile options.
$2683.03 - $16633.87
Specifications
Volume (Metric) 30 μL
Color Clear
Format Rack
Packaging SBS Rack
View More Specs
Products 2
Catalog Number Mfr. No. Sterility Filtered Price Quantity  
Catalog Number Mfr. No. Sterility Filtered Price Quantity  
14-222-368
View Documents
Axygen™
PK384RS
Sterile Non-filtered N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
14-222-367
View Documents
Axygen™
PK384R
Non-sterile Non-filtered
Pack of 3840 Each for $2,683.03
Case of 5 Pack for $13,415.15
Only null left
Description
Description
  • Tip format: 96 tip and 384 tip
  • Tip volume range: 20 μL to 235 μL
  • Tip material: Clear or black polypropylene
  • RNase-/DNase-/human gDNA-free
  • Nonpyrogenic
  • Available options:
    - With or without aerosol-resistant filters
    - Nonsterile or sterile
    - Standard or Axygen™ Maxymum Recovery™ surface
  • Compatible heads: MDT-96 P50, MDT-384-30 μL
  • Rack format: ANSI/SLAS
  • Rack material: Polypropylene
Order Info
Rack: 384 Tips/Rack, 10 Racks/Pack, 5 Packs/Case
Compliance
Axygen products are manufactured in a clean, controlled state of the art facility and certified RNase-, DNase- and endotoxin-safe
Specifications
Specifications
30 μL
Clear
Rack
SBS Rack
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'