Learn More
Innovatek Autoclave Tape
Innovatek Autoclave Tape meets ISO 11140-1:2014 requirements
Supplier: Innovatek 411014424CS
Description
- Made of a saturated crepe paper printed with non lead based ink
- Beige Indicator lines turn dark brown/black once exposed to the steam process
- Coated with an improved synthetic, solvent-free adhesive
- Distinguishes between processed, and unprocessed items
- Does not contain lead and natural rubber latex
- Meets ISO 11140-1:2014 requirement
- Meets US Specification CID A-A-54589A
- Each roll individually wrapped
Specifications
Autoclavable | |
Saturated Crepe Paper | |
Yes | |
24 x 55 mm |
ISO 11140-1:2014, CID A-A-54589A | |
Non-sterile | |
36 Rolls/Cs. | |
Beige to Black Color Change |