missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aushon Chemiluminescent Soluble Human Cytokine Receptor Array Kits
For simultaneous measurement of combinations of soluble cytokine receptors
$847.43 - $1600.71
Specifications
Reactivity | Human |
---|---|
For Use With (Application) | For simultaneous measurement of combinations of soluble Cytokine receptors |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
070011FA
![]() |
Aushon Biosystems Inc
070011FA |
1 Plex |
Each for $847.43
|
|
|||||
07F801FA
![]() |
Aushon Biosystems Inc
07F801FA |
5 Plex |
Each for $1,600.71
|
|
|||||
070061FA
![]() |
Aushon Biosystems Inc
070061FA |
2 Plex |
Each for $1,035.75
|
|
|||||
070781FA
![]() |
Aushon Biosystems Inc
070781FA |
4 Plex |
Each for $1,412.39
|
|
|||||
Description
Using a CCD imaging system and array software, kits allow you to quantify multiple proteins per well in a 96-well plate
Specifications
Human | |
For simultaneous measurement of combinations of soluble Cytokine receptors |