Learn More
Aushon Chemiluminescent Mouse MMP Array Kits DFS Item
For simultaneous measurement of combinations of cytokines
$859.12 - $1050.04
Specifications
For Use With (Application) For simultaneous measurement of combinations of cytokines
View More Specs
Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
200021FA DFS Item
View Documents
Aushon Biosystems Inc
200021FA
1 Plex
Each for $859.12
Only null left
200601FA DFS Item
View Documents
Aushon Biosystems Inc
200601FA
2 Plex
Each for $1,050.04
Only null left
Description
Description
Using a CCD imaging system and array software, kits allow you to quantify multiple proteins per well in a 96-well plate, generating chemokine profiles
Specifications
Specifications
For simultaneous measurement of combinations of cytokines
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'