missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aushon Chemiluminescent Human MMP Array Kits
For simultaneous measurement of matrix metalloproteinase targets
$847.43 - $1224.07
Specifications
Reactivity | Human |
---|---|
For Use With (Application) | For simultaneous measurement of matrix metalloproteinase targets |
Product Type | MMP Array Kits |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
110041FA
![]() |
Aushon Biosystems Inc
110041FA |
1 Plex |
Each for $847.43
|
|
|||||
110021FA
![]() |
Aushon Biosystems Inc
110021FA |
1 Plex |
Each for $847.43
|
|
|||||
110E01FA
![]() |
Aushon Biosystems Inc
110E01FA |
3 Plex |
Each for $1,224.07
|
|
|||||
117001FA
![]() |
Aushon Biosystems Inc
117001FA |
3 Plex |
Each for $1,224.07
|
|
|||||
Description
Using a CCD imaging system and array software, kits allow you to quantify multiple proteins per well in a 96-well plate, generating chemokine profiles
Specifications
Human | |
For simultaneous measurement of matrix metalloproteinase targets | |
MMP Array Kits |