Learn More
Aushon Chemiluminescent Human Inflammation Array Kits DFS Item
For simultaneous measurement of combinations of inflammation targets
Supplier:  Aushon Biosystems Inc 080081FA
Catalog No. 080081FA
Only null left
Explore available promotions
Description
Description
Using a CCD imaging system and array software, kits allow you to quantify multiple proteins per well in a 96-well plate, generating chemokine profiles
Specifications
Specifications
Human
For simultaneous measurement of combinations of inflammation targets
1 Plex
Product Suggestions
Product Suggestions