missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aushon Chemiluminescent Human Cell Adhesion Molecule Array Kits
For simultaneous measurement of combinations of cell adhesion targets
$847.43 - $1412.39
Specifications
Reactivity | Human |
---|---|
For Use With (Application) | For simultaneous measurement of combinations of cell adhesion targets |
Product Type | Human Cell Adhesion Molecule Array Kits |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
080041FA
![]() |
Aushon Biosystems Inc
080041FA |
1 Plex |
Each for $847.43
|
|
|||||
080011FA
![]() |
Aushon Biosystems Inc
080011FA |
4 Plex |
Each for $847.43
|
|
|||||
080E01FA
![]() |
Aushon Biosystems Inc
080E01FA |
3 Plex |
Each for $1,224.07
|
|
|||||
08F001FA
![]() |
Aushon Biosystems Inc
08F001FA |
4 Plex |
Each for $1,412.39
|
|
|||||
Description
Using a CCD imaging system and array software, kits allow you to quantify multiple proteins per well in a 96-well plate, generating chemokine profiles
Specifications
Human | |
For simultaneous measurement of combinations of cell adhesion targets | |
Human Cell Adhesion Molecule Array Kits |