missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aushon Chemiluminescent Human Angiogenesis Array Kits
For simultaneous measurement of combinations of angiogenesis targets
$847.43 - $1412.39
Specifications
Reactivity | Human |
---|---|
For Use With (Application) | For simultaneous measurement of combinations of angiogenesis targets |
Product Type | Angiogenesis Array Kits |
Catalog Number | Mfr. No. | Quantity | Detects | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Detects | Price | Quantity | ||||
090E01FA
![]() |
Aushon Biosystems Inc
090E01FA |
3 Plex | PlGF, TPO, VEGF-R2 |
Each for $1,224.07
|
|
||||
09F001FA
![]() |
Aushon Biosystems Inc
09F001FA |
4 Plex | FGF basic, HGF, PDGF-BB, VEGF |
Each for $1,412.39
|
|
||||
090101FA
![]() |
Aushon Biosystems Inc
090101FA |
1 Plex | VEGF-R3 |
Each for $847.43
|
|
||||
090041FA
![]() |
Aushon Biosystems Inc
090041FA |
1 Plex | PDGF-AA |
Each for $847.43
|
|
||||
090021FA
![]() |
Aushon Biosystems Inc
090021FA |
1 Plex | HB-EGF |
Each for $847.43
|
|
||||
090081FA
![]() |
Aushon Biosystems Inc
090081FA |
1 Plex | VEGF-R1 |
Each for $847.43
|
|
||||
Description
Using a CCD imaging system and array software, kits allow you to quantify multiple proteins per well in a 96-well plate, generating chemokine profiles
Specifications
Human | |
For simultaneous measurement of combinations of angiogenesis targets | |
Angiogenesis Array Kits |