Learn More
Astoria-Pacific Micro Flow Astoria 2 Analyzer System Replacement Parts and Accessories 4 DFS Item
$103.72 - $103.72
Specifications
Type Micropump Tube
View More Specs
Products 4
Catalog Number Mfr. No. Color Flow Rate Price Quantity  
Catalog Number Mfr. No. Color Flow Rate Price Quantity  
303-1035P01 DFS Item
View Documents
Selkirk Scientific
3031035P01
Orange 287 μL/min.
Pack of 12 Each for $103.72
Only null left
303-1020P01 DFS Item
View Documents
Selkirk Scientific
3031020P01
Orange/Yellow 118 μL/min.
Pack of 12 Each for $103.72
Only null left
303-1051P01 DFS Item
View Documents
Selkirk Scientific
3031051P01
Gray 568 μL/min.
Pack of 12 Each for $103.72
Only null left
303-1030P01 DFS Item
View Documents
Selkirk Scientific
3031030P01
Black 226 μL/min.
Pack of 12 Each for $103.72
Only null left
Description
Description
Specifications
Specifications
Micropump Tube
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'