missing translation for 'onlineSavingsMsg'
Learn More

Medicom™ AssureTouch™ Agile™ Nitrile Medical Examination Gloves with Textured Fingertips

AssureTouch™ Agile™ Nitrile Medical Examination Gloves with Textured Fingertips. This nitrile glove offers optimal puncture and tear resistance without compromising dexterity or tactility. Textured fingertips ensure a secure grip, even when wet. Chemo tested.

$13.83 - $13.83

Specifications

Chlorinated No
Accelerator Free No
Applications Healthcare, Industrial, Laboratory
AQL 1.5
Color Blue
View More Specs

Products 5
Catalog Number Mfr. No. Size Price Quantity  
Catalog Number Mfr. No. Size Price Quantity  
17101083
View Documents
Medicom™
9998A
X-Small
Pack of 100 Each for $13.83
Case of 10 Pack for $138.30
Only null left
 
17101084 Edge
View Documents
Medicom™
9998B
Small
Pack of 100 Each for $13.83
Case of 10 Pack for $138.30
Only null left
Edge
 
17101085
View Documents
Medicom™
9998C
Medium
Pack of 100 Each for $13.83
Case of 10 Pack for $138.30
Only null left
 
17101086
View Documents
Medicom™
9998D
Large
Pack of 100 Each for $13.83
Case of 10 Pack for $138.30
Only null left
 
17101087
View Documents
Medicom™
9998E
X-Large
Pack of 100 Each for $13.83
Case of 10 Pack for $138.30
Only null left
 
Description

Description

  • These nitrile gloves offer superior puncture, tear and chemical resistance without compromising dexterity or tactility.
  • Textured fingertips ensure a secure grip, even when wet.
  • Chemo tested.
Specifications

Specifications

No
Healthcare, Industrial, Laboratory
Blue
Fingertip Textured
Powder Free
Yes
No
1.5
Single-use
Yes
Non-sterile
Beaded
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'