Learn More
United Scientific™ Aspirator Bottles
Heavy duty for safe storage.
$109.00 - $247.37
Specifications
Color | White |
---|---|
Material | Polypropylene |
Sterility | Non-sterile |
Autoclavable | Autoclavable |
Shape | Cylindrical |
Description
- These lightweight, autoclavable aspirator bottles are made from polypropylene and can be used for dispensing distilled water and other laboratory solutions.
- The leak-proof spigot screwed to the bottle requires only a half turn to open or close.
- Polyethylene cap and stopcock should be removed before autoclaving.
Specifications
White | |
Non-sterile | |
Cylindrical |
Polypropylene | |
Autoclavable | |
Aspirator Bottle |
Warning: Cancer and reproductive harm