missing translation for 'onlineSavingsMsg'
Learn More

Ascorbic Acid, Crystalline Powder, USP, 99-100.5%, Spectrum™ Chemical

AS105, 50-81-7, C6H8O6

$205.91 - $40079.03

Chemical Identifiers

CAS 50-81-7
Molecular Formula C6H8O6
Molecular Weight (g/mol) 176.12
MDL Number MFCD00064328
InChI Key CIWBSHSKHKDKBQ-DOMZIZNONA-N
IUPAC Name (5R)-5-[(1S)-1,2-dihydroxyethyl]-3,4-dihydroxy-2,5-dihydrofuran-2-one
SMILES OC[C@H](O)[C@H]1OC(=O)C(O)=C1O
View More Specs

Products 7
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
18600770
View Documents
Spectrum Chemical Mfg Cor
AS10525GM
25 g Amber Glass Bottle
Each for $205.91
Only null left
 
18600771
View Documents
Spectrum Chemical Mfg Cor
AS105100GM
100 g Amber Glass Bottle
Each for $246.86
Only null left
 
18600766
View Documents
Spectrum Chemical Mfg Cor
AS105500GM
500 g Amber Glass Bottle
Each for $656.20
Only null left
 
18600767
View Documents
Spectrum Chemical Mfg Cor
AS1051KG
1 kg Amber Glass Bottle
Each for $1,113.49
Only null left
 
18600768
View Documents
Spectrum Chemical Mfg Cor
AS1055KG
5 kg Poly Pail
Each for $4,172.17
Only null left
 
18600769
View Documents
Spectrum Chemical Mfg Cor
AS10512KG
12 kg Poly Pail
Each for $7,560.86
Only null left
 
18600772
View Documents
Spectrum Chemical Mfg Cor
AS10550KGBL
50 kg Fiber Drum N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Spectrum™ Chemical Ascorbic Acid, Crystalline Powder, USP , Vitamin C, is a dietary supplement used to treat and prevent vitamin C deficiency. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Specifications

Chemical Identifiers

50-81-7
176.12
CIWBSHSKHKDKBQ-DOMZIZNONA-N
OC[C@H](O)[C@H]1OC(=O)C(O)=C1O
C6H8O6
MFCD00064328
(5R)-5-[(1S)-1,2-dihydroxyethyl]-3,4-dihydroxy-2,5-dihydrofuran-2-one

Specifications

50-81-7
0.1%
C6H8O6
25 g
CIWBSHSKHKDKBQ-DOMZIZNONA-N
(5R)-5-[(1S)-1,2-dihydroxyethyl]-3,4-dihydroxy-2,5-dihydrofuran-2-one
99 to 100.5%
100%
Amber Glass Bottle
MFCD00064328
+20.5° to +21.5°
OC[C@H](O)[C@H]1OC(=O)C(O)=C1O
176.12
USP
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'