missing translation for 'onlineSavingsMsg'
Learn More

Arsenazo III 95.0+%, TCI America™

[Spectrophotometric reagent for U, Th, Zr and other metals, Indicator for the precipitation titration of SO4 with Ba]

$117.43 - $346.71

Chemical Identifiers

CAS 1668-00-4
Molecular Formula C22H18As2N4O14S2
Molecular Weight (g/mol) 776.363
MDL Number MFCD00036695
InChI Key TVMZRHVOFZTNET-RIRMOVKSSA-N
Synonym arsenazo iii, arsenazo iii sodium salt, 2,2'-1, 8-dihydroxy-3,6-disulfo-2, 7-naphthalene-bis azo dibenzenearsonic acid, 2,7-bis 2-arsonophenylazo-1, 8-dihydroxy-3, 6-naphthalene disulfonic acid, 2,7-bis 2-arsonophenylazo-1,8-dihydroxynaphthalene-3,6-disulfonic acid, 2,7-bis 2-arsonophenylazo chromotropic acid, 2,2'-1,8-dihydroxy-3,6-disulfonaphthylene-2,7-bisazo bisbenzenearsonic acid, 3,6-bis 2-2-arsonophenyl diazen-1-yl-4,5-dihydroxynaphthalene-2,7-disulfonic acid, 3,6-bis 2-2-arsonophenyl diazenyl-4,5-dihydroxy-2,7-naphthalenedisulfonic acid, 3,6-bis 2-2-arsonophenyl hydrazin-1-ylidene-4,5-dioxonaphthalene-2,7-disulfonic acid
PubChem CID 9810878
IUPAC Name (3Z,6E)-3,6-bis[(2-arsonophenyl)hydrazinylidene]-4,5-dioxonaphthalene-2,7-disulfonic acid
SMILES C1=CC=C(C(=C1)NN=C2C(=CC3=C(C2=O)C(=O)C(=NNC4=CC=CC=C4[As](=O)(O)O)C(=C3)S(=O)(=O)O)S(=O)(=O)O)[As](=O)(O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
A50091G
View Documents
TCI America
A50091G
1 g
Each for $117.43
Only null left
 
A50095G
View Documents
TCI America
A50095G
5 g
Each for $346.71
Only null left
 
Description

Description

Specifications

Chemical Identifiers

1668-00-4
776.363
TVMZRHVOFZTNET-RIRMOVKSSA-N
9810878
C1=CC=C(C(=C1)NN=C2C(=CC3=C(C2=O)C(=O)C(=NNC4=CC=CC=C4[As](=O)(O)O)C(=C3)S(=O)(=O)O)S(=O)(=O)O)[As](=O)(O)O
C22H18As2N4O14S2
MFCD00036695
arsenazo iii, arsenazo iii sodium salt, 2,2'-1, 8-dihydroxy-3,6-disulfo-2, 7-naphthalene-bis azo dibenzenearsonic acid, 2,7-bis 2-arsonophenylazo-1, 8-dihydroxy-3, 6-naphthalene disulfonic acid, 2,7-bis 2-arsonophenylazo-1,8-dihydroxynaphthalene-3,6-disulfonic acid, 2,7-bis 2-arsonophenylazo chromotropic acid, 2,2'-1,8-dihydroxy-3,6-disulfonaphthylene-2,7-bisazo bisbenzenearsonic acid, 3,6-bis 2-2-arsonophenyl diazen-1-yl-4,5-dihydroxynaphthalene-2,7-disulfonic acid, 3,6-bis 2-2-arsonophenyl diazenyl-4,5-dihydroxy-2,7-naphthalenedisulfonic acid, 3,6-bis 2-2-arsonophenyl hydrazin-1-ylidene-4,5-dioxonaphthalene-2,7-disulfonic acid
(3Z,6E)-3,6-bis[(2-arsonophenyl)hydrazinylidene]-4,5-dioxonaphthalene-2,7-disulfonic acid

Specifications

1668-00-4
MFCD00036695
3465
TVMZRHVOFZTNET-RIRMOVKSSA-N
(3Z,6E)-3,6-bis[(2-arsonophenyl)hydrazinylidene]-4,5-dioxonaphthalene-2,7-disulfonic acid
9810878
≥95.0% (T)
C22H18As2N4O14S2
1 g
arsenazo iii, arsenazo iii sodium salt, 2,2'-1, 8-dihydroxy-3,6-disulfo-2, 7-naphthalene-bis azo dibenzenearsonic acid, 2,7-bis 2-arsonophenylazo-1, 8-dihydroxy-3, 6-naphthalene disulfonic acid, 2,7-bis 2-arsonophenylazo-1,8-dihydroxynaphthalene-3,6-disulfonic acid, 2,7-bis 2-arsonophenylazo chromotropic acid, 2,2'-1,8-dihydroxy-3,6-disulfonaphthylene-2,7-bisazo bisbenzenearsonic acid, 3,6-bis 2-2-arsonophenyl diazen-1-yl-4,5-dihydroxynaphthalene-2,7-disulfonic acid, 3,6-bis 2-2-arsonophenyl diazenyl-4,5-dihydroxy-2,7-naphthalenedisulfonic acid, 3,6-bis 2-2-arsonophenyl hydrazin-1-ylidene-4,5-dioxonaphthalene-2,7-disulfonic acid
C1=CC=C(C(=C1)NN=C2C(=CC3=C(C2=O)C(=O)C(=NNC4=CC=CC=C4[As](=O)(O)O)C(=C3)S(=O)(=O)O)S(=O)(=O)O)[As](=O)(O)O
776.363
776.36
Arsenazo III [Spectrophotometric reagent for U, Th, Zr and other metals, Indicator for the precipita
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'