missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Arbutin 95.0+%, TCI America™
$102.52 - $288.94
Chemical Identifiers
CAS | 497-76-7 |
---|---|
Molecular Formula | C12H16O7 |
Molecular Weight (g/mol) | 272.25 |
MDL Number | MFCD00016915 |
InChI Key | BJRNKVDFDLYUGJ-UHFFFAOYNA-N |
Synonym | arbutin, uvasol, 4-hydroxyphenyl beta-d-glucopyranoside, ursin, beta-arbutin, arbutoside, arbutine, arbutyne, ursi, p-arbutin |
PubChem CID | 440936 |
ChEBI | CHEBI:18305 |
IUPAC Name | 2-(hydroxymethyl)-6-(4-hydroxyphenoxy)oxane-3,4,5-triol |
SMILES | OCC1OC(OC2=CC=C(O)C=C2)C(O)C(O)C1O |
Chemical Identifiers
497-76-7 | |
272.25 | |
BJRNKVDFDLYUGJ-UHFFFAOYNA-N | |
440936 | |
2-(hydroxymethyl)-6-(4-hydroxyphenoxy)oxane-3,4,5-triol |
C12H16O7 | |
MFCD00016915 | |
arbutin, uvasol, 4-hydroxyphenyl beta-d-glucopyranoside, ursin, beta-arbutin, arbutoside, arbutine, arbutyne, ursi, p-arbutin | |
CHEBI:18305 | |
OCC1OC(OC2=CC=C(O)C=C2)C(O)C(O)C1O |
Specifications
497-76-7 | |
White | |
MFCD00016915 | |
arbutin, uvasol, 4-hydroxyphenyl beta-d-glucopyranoside, ursin, beta-arbutin, arbutoside, arbutine, arbutyne, ursi, p-arbutin | |
OCC1OC(OC2=CC=C(O)C=C2)C(O)C(O)C1O | |
272.25 | |
CHEBI:18305 | |
≥95.0% (HPLC) | |
Arbutin |
198°C | |
C12H16O7 | |
5 g | |
BJRNKVDFDLYUGJ-UHFFFAOYNA-N | |
2-(hydroxymethyl)-6-(4-hydroxyphenoxy)oxane-3,4,5-triol | |
440936 | |
272.25 | |
Crystalline Powder |
Safety and Handling
RTECSNumber : CE8863000
TSCA : No