missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ MicroAmp™ 48-Well Base Adaptor DFS Item

For use with 48-well plate format

Supplier:  Applied Biosystems™ 4375284

Catalog No. 4375284


Only null left
Explore available promotions

Includes: 5 MicroAmp 48-Well Base Adaptors.

Description

Description

Specifically designed to fit onto MicroAmp 96-Well Support Base.

See user's manual or package insert for limited label license and trademark information.

PCR, PCR and Real-Time PCR, Real Time PCR (qPCR)

Order Info

Shipping Condition: Room temperature

Specifications

Specifications

Store at room temperature.
StepOne™,
5 Adaptors
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

For Research Use Only. Not for use in diagnostic procedures.