missing translation for 'onlineSavingsMsg'
Learn More

Antimony(Iii) Chloride, puriss. p.a.ACS Reagent, ≥99.0% (RT), Honeywell Fluka™

Puriss. p.a., ACS reagent,≥99.0% (RT)

$1391.18 - $1391.18

Chemical Identifiers

CAS 10025-91-9
Molecular Formula Cl3Sb
Molecular Weight (g/mol) 228.11
MDL Number MFCD00011212
InChI Key FAPDDOBMIUGHIN-UHFFFAOYSA-K
Synonym antimony trichloride, trichlorostibine, antimony chloride, antimony iii chloride, stibine, trichloro, antimontrichlorid, antimonous chloride, butter of antimony, antimony butter, caustic antimony
PubChem CID 24814
ChEBI CHEBI:74856
IUPAC Name trichlorostibane
SMILES [Cl-].[Cl-].[Cl-].[Sb+3]
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
60044658
View Documents
Honeywell Chemicals
10774500G
500 g Poly Bottle
Each for $1,391.18
Only null left
 
Description

Description

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications

Chemical Identifiers

10025-91-9
228.11
FAPDDOBMIUGHIN-UHFFFAOYSA-K
24814
trichlorostibane
Cl3Sb
MFCD00011212
antimony trichloride, trichlorostibine, antimony chloride, antimony iii chloride, stibine, trichloro, antimontrichlorid, antimonous chloride, butter of antimony, antimony butter, caustic antimony
CHEBI:74856
[Cl-].[Cl-].[Cl-].[Sb+3]

Specifications

283°C
10025-91-9
500 g
SbCl3
UN1733
FAPDDOBMIUGHIN-UHFFFAOYSA-K
trichlorostibane
24814
228.12g/mol
ACS/puriss. p.a.
Crystals/Powder
3.140g/cu.cm; Vapor Density: 7.9 (vs air)
73.4°C
White
Cl3Sb
MFCD00011212
antimony trichloride, trichlorostibine, antimony chloride, antimony iii chloride, stibine, trichloro, antimontrichlorid, antimonous chloride, butter of antimony, antimony butter, caustic antimony
[Cl-].[Cl-].[Cl-].[Sb+3]
228.11
CHEBI:74856
≥99.0% (RT)
Poly Bottle
1mmHg (49°C)
Antimony(Iii) Chloride
Videos
Safety and Handling

Safety and Handling

P273-P280-P303 + P361 + P353-P304 + P340 + P310-P305 + P351 + P338-P501

H314-H411

DOTInformation : 8/PGII

EINECSNumber : 233-047-2

RTECSNumber : CC4900000

SDS
missing translation for 'documents'

missing translation for 'documents'