missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TIMELESS interacting protein, Mouse, Purified MaxPab™ Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a full-length human FLJ20516 protein.
Supplier: Abnova Corporation H00054962B01P
Description
Sequence: MLEPQENGVIDLPDYEHVEDETFPPFPPPASPERQDGEGTEPDEESGNGAPVPVPPKRTVKRNIPKLDAQRLISERGLPALRHVFDKAKFKGKGHEAEDLKMLIRHMEHWAHRLFPKLQFEDFIDRVEYLGSKKEVQTCLKRIRLDLPILHEDFVSNNDEVAENNEHDVTSTELDPFLTNLSESEMFASELSRSLTEEQQQRIERNKQLALERRQAKLLSNSQTLGNDMLMNTPRAHTVEEVNTDEDQKEESNGLNEDILDNPCNDAIANTLNEEETLLDQSFKNVQQQLDATSRNITEARSpecifications
TIMELESS interacting protein | |
Polyclonal | |
Mouse polyclonal antibody raised against a full-length human FLJ20516 protein. | |
TIPIN | |
FLJ20516 | |
Mouse | |
Affinity Purified | |
RUO | |
54962 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG |
Western Blot | |
Unconjugated | |
PBS with no preservative; pH 7.4 | |
BC000870.1 | |
TIPIN | |
FLJ20516 (AAH00870.1, 1 a.a. ∼ 301 a.a) full-length human protein. | |
50 μg | |
Primary | |
Human | |
Antibody |