Learn More
KNF™ FLEXPAK Anti-static Block Bottom Bags
Clear and clean polyethylene bags with a 4 mil thickness.
Supplier:  KNF™ FLEXPAK 020507448
 View more versions of this product
Catalog No. 17020219
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Explore available promotions
Description
Description
  • Clear, anti-static plastic
  • Square bottom
Specifications
Specifications
Not Autoclavable
Clear
4 mil
121.9 cm
Level 200 Clean
Polyethylene
48 x 48 in.
203.2 cm
Product Suggestions
Product Suggestions