Learn More
Anti-Sialyl Lewis A Monoclonal Antibody (2D3) , TCI America™
Anti-Sialyl Lewis A Monoclonal Antibody (2D3), TCI America™
Supplier:  TCI America A25091VIAL
Catalog No. A2509VIAL
Only null left
Explore available promotions
Description
Description
Specifications
Specifications
Sialyl Lewis A
Monoclonal
Mouse
Primary
Purified
Immunohistochemistry, Flow Cytometry, ELISA, TLC-Immunostaining, Western Blot, Inhibition Assays
2D3
1 Vial
Store at -20°C to -80°C (preferably at -80°C). It is not recommended storing at 4°C. Avoid repeated freezing and thawing.
IgM κ
Product Suggestions
Product Suggestions