missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Anti-Lewis X Monoclonal Antibody , TCI America™
Anti-Lewis X Monoclonal Antibody, TCI America™
Supplier: TCI America A25781VIAL
Specifications
Lewis X | |
Monoclonal | |
Mouse | |
Primary | |
Purified |
ELISA, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Western Blot, TLC-Immunostaining | |
73-30 | |
Salmonella Minnesota coated with Lewis X-active glycolipids | |
Store at -20°C to -80°C (preferably at -80°C). It is not recommended storing at 4°C. Avoid repeated freezing and thawing. | |
IgM κ |