missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Anti-Keratan Sulfate Monoclonal Antibody (R-10G) , TCI America™
Anti-Keratan Sulfate Monoclonal Antibody (R-10G), TCI America™
Supplier: TCI America A29681VIAL
Specifications
Keratan Sulfate | |
Monoclonal | |
Mouse | |
1 Vial | |
Store at -20°C to -80°C (preferably at -80°C). It is not recommended storing at 4°C. Avoid repeated freezing and thawing. | |
IgG1 |
Flow Cytometry, Immunoprecipitation, Immunoaffinity Chromatography, ELISA, Western Blot, Immunohistochemistry | |
R-10G | |
Human iPS cell (Tic) | |
Primary | |
Purified |