missing translation for 'onlineSavingsMsg'
Learn More
Learn More
JMJD3 (NT) Rabbit anti-Human, Mouse, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma 071533
Description
Use Anti-JMJD3 (N-terminus) Antibody (Rabbit Polyclonal Antibody) validated in immunohistochemistry, immunocytochemistry, ELISA, western blotting to detect JMJD3 (N-terminus) also known as JmjC domain-containing protein 3.Specifications
JMJD3 (NT) | |
Polyclonal | |
Purified polyclonal antibody supplied in PBS with 0.09% (W/V) sodium azide. | |
Rabbit | |
Protein G chromatography | |
Primary | |
Human, Mouse | |
Purified |
ELISA, Immunocytochemistry, Immunohistochemistry, Western Blot | |
Unconjugated | |
EC 1.14.11; KDM6B ; KIAA0346 | |
This antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide selected from the N-terminal region of human JMJD3. | |
100 μg | |
NP_001073893 | |
2-8°C for 12 months Handling Recommendations: Upon receipt and prior to removing the cap centrifuge the vial to pellet the solution. Avoid repeated freeze/thaw cycles which may damage IgG and affect product performance. |
For Research Use Only