Learn More
Anti-GQ1b Monoclonal Antibody , TCI America™
Anti-GQ1b Monoclonal Antibody, TCI America™
Supplier:  TCI America A26621VIAL
Catalog No. A2662VIAL
Only null left
Explore available promotions
Description
Description
Specifications
Specifications
GQ1b
Monoclonal
Mouse
1 Vial
Store at -20°C to -80°C (preferably at -80°C). It is not recommended storing at 4°C. Avoid repeated freezing and thawing.
IgM κ
TLC-Immunostaining, ELISA, Immunohistochemistry
GMR13
Purified ganglioside GQ1 adsorbed to Salmonella Minnesota
Primary
Purified
Product Suggestions
Product Suggestions