missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Anti-GM1 Monoclonal Antibody , TCI America™
Anti-GM1 Monoclonal Antibody, TCI America™
Supplier: TCI America A25051VIAL
Specifications
GM-1 | |
Monoclonal | |
Mouse | |
1 Vial | |
Store at -20°C to -80°C (preferably at -80°C). It is not recommended storing at 4°C. Avoid repeated freezing and thawing. |
TLC-Immunostaining, Flow Cytometry, ELISA, Immunocytochemistry, Immunohistochemistry | |
GMB16 | |
Purified ganglioside GM1 | |
Primary | |
Purified |