Learn More
Anti-GD1b Monoclonal Antibody , TCI America™
Anti-GD1b Monoclonal Antibody, TCI America™
Supplier:  TCI America A25081VIAL
Catalog No. A2508VIAL
Only null left
Explore available promotions
Description
Description
Specifications
Specifications
GD1b
Monoclonal
Mouse
1 Vial
Store at -20°C to -80°C (preferably at -80°C). It is not recommended storing at 4°C. Avoid repeated freezing and thawing.
IgG3 κ
TLC-Immunostaining, Flow Cytometry, ELISA, Immunocytochemistry, Immunohistochemistry
GGR12
Purified ganglioside GD1b adsorbed to Salmonella Minnesota
Primary
Purified
Product Suggestions
Product Suggestions