missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FOXO1A, Mouse, Clone: 4D9, Abnova™
Mouse monoclonal antibody raised against a partial recombinant FOXO1A.
Supplier: Abnova Corporation H00002308M05
Description
This gene belongs to the forkhead family of transcription factors which are characterized by a distinct forkhead domain. The specific function of this gene has not yet been determined; however, it may play a role in myogenic growth and differentiation. Translocation of this gene with PAX3 has been associated with alveolar rhabdomyosarcoma. [provided by RefSeq
Sequence: TQVKTPVQVPLPHPMQMSALGGYSSVSSCNGYGRMGLLHQEKLPSDLDGMFIERLDCDMESIIRNDLMDGDTLDFNFDNVLPNQSFPHSVKTTTHSWVSGSpecifications
FOXO1A | |
Monoclonal | |
Unconjugated | |
PBS with no preservative; pH 7.4 | |
NM_002015 | |
FOXO1 | |
FOXO1A (NP_002006, 556 a.a. ∼ 655 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2a κ |
ELISA, Western Blot | |
4D9 | |
Mouse monoclonal antibody raised against a partial recombinant FOXO1A. | |
FOXO1 | |
FKH1/FKHR/FOXO1A | |
Mouse | |
Affinity chromatography | |
RUO | |
2308 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
Liquid |