missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FLI1, Mouse, Clone: 2F3, Abnova™
Mouse monoclonal antibody raised against a full-length recombinant FLI1.
Supplier: Abnova Corporation H00002313M09
Description
Sequence: MDGTIKEALSVVSDGQSLFDSAYGAAAHLPKADMAASGSPDYGQPHKINPLPPQQEWISQPVRVNVKREYDHMNGSRESPVDCSVSKCSKLVGGGESNPMNYNSYMDEKNGPPPPNMTTNERRVIVPADPTLWTQEHVRQWLEWAIKEYSLMEIDTSFFQNMDGKELCKMNKEDFLRATTLYNTEVLLSHLSYLRESSLLAYNTTSHTDQSSRLSVKEDPSYDSVRRGAWGNNMNSGLNKSPPLGGAQTISKNTEQRPQPDPYQILGPTSSRLANPGSGQIQLWQFLLELLSDSANASCITWEGTNGEFKMTDPDEVARRWGERISKPNMNYDKLSRALRYYYDKNIMTKVHGKRYAYKFDFHGIAQALQPHPTESSMYKYPSDISYMPSYHAHQQKVNFVPPHPSSMPVTSSSFFGAASQYWTSPTGGIYPNPNVPRHPNTHVPSHLGSYYSpecifications
FLI1 | |
Monoclonal | |
Unconjugated | |
PBS with no preservative; pH 7.4 | |
BC001670 | |
FLI1 | |
FLI1 (AAH01670, 1 a.a. ∼ 452 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2a λ |
ELISA, Immunofluorescence, Western Blot | |
2F3 | |
Mouse monoclonal antibody raised against a full length recombinant FLI1. | |
FLI1 | |
EWSR2/SIC-1 | |
Mouse | |
Affinity chromatography | |
RUO | |
2313 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
Liquid |