Learn More
FGF10, Mouse, Clone: 3C7, Abnova™
Mouse monoclonal antibody raised against a partial recombinant FGF10.
Supplier: Abnova Corporation H00002255M05
Description
The protein encoded by this gene is a member of the fibroblast growth factor (FGF) family. FGF family members possess broad mitogenic and cell survival activities, and are involved in a variety of biological processes, including embryonic development, cell growth, morphogenesis, tissue repair, tumor growth and invasion. This protein exhibits mitogenic activity for keratinizing epidermal cells, but essentially no activity for fibroblasts, which is similar to the biological activity of FGF7. Studies of the mouse homolog of suggested that this gene is required for embryonic epidermal morphogenesis including brain development, lung morphogenesis, and initiation of lim bud formation. This gene is also implicated to be a primary factor in the process of wound healing. [provided by RefSeq
Sequence: QALGQDMVSPEATNSSSSSFSSPSSAGRHVRSYNHLQGDVRWRKLFSFTKYFLKIEKNGKVSGTKKENCPYSILEITSVEIGVVAVKAINSNYYLAMNKKSpecifications
FGF10 | |
Monoclonal | |
Unconjugated | |
PBS with no preservative; pH 7.4 | |
NM_004465 | |
Mouse | |
Affinity chromatography | |
RUO | |
2255 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
Liquid |
ELISA, In situ PLA, Western Blot | |
3C7 | |
Mouse monoclonal antibody raised against a partial recombinant FGF10. | |
FGF10 | |
FGF10 | |
FGF10 (NP_004456, 38 a.a. ∼ 137 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2a κ |