Learn More
FANCC, Mouse, Clone: 6E7, Abnova™
Mouse monoclonal antibody raised against a partial recombinant FANCC.
Supplier: Abnova Corporation H00002176M01
Description
The Fanconi anemia complementation group (FANC) currently includes FANCA, FANCB, FANCC, FANCD1 (also called BRCA2), FANCD2, FANCE, FANCF, FANCG, FANCI, FANCJ (also called BRIP1), FANCL, FANCM and FANCN (also called PALB2). The previously defined group FANCH is the same as FANCA. Fanconi anemia is a genetically heterogeneous recessive disorder characterized by cytogenetic instability, hypersensitivity to DNA crosslinking agents, increased chromosomal breakage, and defective DNA repair. The members of the Fanconi anemia complementation group do not share sequence similarity; they are related by their assembly into a common nuclear protein complex. This gene encodes the protein for complementation group C. [provided by RefSeq
Sequence: MAQDSVDLSCDYQFWMQKLSVWDQASTLETQQDTCLHVAQFQEFLRKMYEALKEMDSNTVIERFPTIGQLLAKACWNPFILAYDESQKILIWCLCCLINKSpecifications
FANCC | |
Monoclonal | |
Unconjugated | |
PBS with no preservative; pH 7.4 | |
NM_000136 | |
FANCC | |
FANCC (NP_000127, 1 a.a. ∼ 100 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2b κ |
ELISA, Immunofluorescence, Western Blot | |
6E7 | |
Mouse monoclonal antibody raised against a partial recombinant FANCC. | |
FANCC | |
FA3/FAC/FACC/FLJ14675 | |
Mouse | |
Affinity chromatography | |
RUO | |
2176 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
Liquid |