missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Anti-Endo-M Polyclonal Antibody , TCI America™
Anti-Endo-M Polyclonal Antibody, TCI America™
Supplier: TCI America A29581VIAL
Specifications
Endo-M | |
Polyclonal | |
endo-β-N-Acetylglucosaminidase (Endo-M) from Mucor hiemalis | |
Primary | |
Purified |
ELISA, Western Blot | |
Rabbit | |
1 Vial | |
Store at -20°C to -80°C (preferably at -80°C). It is not recommended storing at 4°C. Avoid repeated freezing and thawing. | |
IgG |