missing translation for 'onlineSavingsMsg'
Learn More

Anhydrous Citric Acid, Granular, USP, 99.5-100.5%, Spectrum™ Chemical

CI133, 77-92-9, C6H8O7

$549.80 - $6675.23

Chemical Identifiers

CAS 77-92-9
Molecular Formula C6H8O7
Molecular Weight (g/mol) 192.12
MDL Number MFCD00011669
InChI Key KRKNYBCHXYNGOX-UHFFFAOYSA-N
IUPAC Name 2-hydroxypropane-1,2,3-tricarboxylic acid
SMILES OC(=O)CC(O)(CC(O)=O)C(O)=O
View More Specs

Products 5
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
18600702
View Documents
Spectrum Chemical Mfg Cor
CI133500GM
500 g Poly Bottle
Each for $549.80
Only null left
 
18600703
View Documents
Spectrum Chemical Mfg Cor
CI13325KG
2.5 kg Poly Bottle
Each for $1,538.13
Only null left
 
18600704
View Documents
Spectrum Chemical Mfg Cor
CI13312KG
12 kg Poly Pail
Each for $3,608.80
Only null left
 
18600706
View Documents
Spectrum Chemical Mfg Cor
CI13325KGBL
25 kg Fiber Drum
Each for $6,429.30
Only null left
 
18600705
View Documents
Spectrum Chemical Mfg Cor
CI13345KGBL
45 kg Fiber Drum
Each for $6,675.23
Only null left
 
Description

Description

Spectrum™ Chemical Anhydrous Citric Acid, Granular, USP belongs to a class of drugs known as urinary alkalinizers that are used to treat certain metabolic problems (acidosis) caused by kidney disease. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Specifications

Chemical Identifiers

77-92-9
192.12
KRKNYBCHXYNGOX-UHFFFAOYSA-N
OC(=O)CC(O)(CC(O)=O)C(O)=O
C6H8O7
MFCD00011669
2-hydroxypropane-1,2,3-tricarboxylic acid

Specifications

77-92-9
C6H8O7
KRKNYBCHXYNGOX-UHFFFAOYSA-N
2-hydroxypropane-1,2,3-tricarboxylic acid
99.5 to 100.5%
500 g
Poly Bottle
100%
MFCD00011669
OC(=O)CC(O)(CC(O)=O)C(O)=O
192.12
USP
0.1%
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'