missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium Thiocyanate 98.0+%, TCI America™
$48.46 - $132.35
Chemical Identifiers
CAS | 1762-95-4 |
---|---|
Molecular Formula | CH4N2S |
Molecular Weight (g/mol) | 76.117 |
MDL Number | MFCD00011428 |
InChI Key | SOIFLUNRINLCBN-UHFFFAOYSA-N |
Synonym | ammonium thiocyanate, ammoniumrhodanid, thiocyanic acid, ammonium salt, ammonium isothiocyanate, ammonium rhodanate, weedazol tl, trans-aid, ammonium sulfocyanate, ammonium sulfocyanide, rhodanine, ammonium salt |
PubChem CID | 15666 |
IUPAC Name | azanium;thiocyanate |
SMILES | C(#N)[S-].[NH4+] |
Chemical Identifiers
1762-95-4 | |
76.117 | |
SOIFLUNRINLCBN-UHFFFAOYSA-N | |
15666 | |
C(#N)[S-].[NH4+] |
CH4N2S | |
MFCD00011428 | |
ammonium thiocyanate, ammoniumrhodanid, thiocyanic acid, ammonium salt, ammonium isothiocyanate, ammonium rhodanate, weedazol tl, trans-aid, ammonium sulfocyanate, ammonium sulfocyanide, rhodanine, ammonium salt | |
azanium;thiocyanate |
Specifications
1762-95-4 | |
White | |
CH4N2S | |
25 g | |
SOIFLUNRINLCBN-UHFFFAOYSA-N | |
azanium;thiocyanate | |
15666 | |
≥98.0% (T) | |
Ammonium Thiocyanate |
150°C | |
4.0 - 5.5 | |
MFCD00011428 | |
ammonium thiocyanate, ammoniumrhodanid, thiocyanic acid, ammonium salt, ammonium isothiocyanate, ammonium rhodanate, weedazol tl, trans-aid, ammonium sulfocyanate, ammonium sulfocyanide, rhodanine, ammonium salt | |
C(#N)[S-].[NH4+] | |
76.117 | |
76.12 | |
Crystalline Powder |
Safety and Handling
EINECSNumber : (1)-0142
RTECSNumber : XK7875000
TSCA : Yes