missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Standard, 1000 ppm NH4+ (776.5 ppm N), Ricca Chemical

NH4Cl in Water

$221.94 - $368.44

Chemical Identifiers

CAS 12125-02-9
Molecular Formula ClH4N
Molecular Weight (g/mol) 53.49
MDL Number MFCD00011420
InChI Key NLXLAEXVIDQMFP-UHFFFAOYSA-N
PubChem CID 25517
ChEBI CHEBI:31206
IUPAC Name amine hydrochloride
SMILES N.Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
693-16
View Documents
Ricca Chemical Company
69316
500 mL Natural Poly Bottle
Each for $221.94
Only null left
 
693-32
SDS View Documents
Ricca Chemical Company
69332
1 L Natural Poly Bottle
Each for $368.44
Only null left
 
Description

Description

Specifications

Chemical Identifiers

12125-02-9
53.49
NLXLAEXVIDQMFP-UHFFFAOYSA-N
CHEBI:31206
N.Cl
ClH4N
MFCD00011420
25517
amine hydrochloride

Specifications

Natural Poly Bottle
98.0,0.29
1 mL = 1 mg NH
ClH4N
Colorless
NLXLAEXVIDQMFP-UHFFFAOYSA-N
amine hydrochloride
500 mL
CHEBI:31206
12125-02-9,7732-18-5
100.0,0.3
Liquid
MFCD00011420
100°C
N.Cl
53.49
25517
Ammonium Standard
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'