missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium Hydroxide, Reagent, ACS, 28-30%, Spectrum™ Chemical
A1195, 1336-21-6, NH4OH
Supplier: Spectrum Chemical Mfg Cor A119525LTPVCS6
Description
Spectrum™ Chemical Ammonium Hydroxide, Reagent, ACS is a solution of amonia in water. Aqueous amonia, as it′s otherwise known is used as a complexant and base in qualitative inorganic analysis. It becomes a deep blue color with copper solutions. Also used to dissolve silver oxide residues, such as the ones formed from Tollens′ reagent. It is also commmonly used in numerous household cleaning agents, such as window cleaning formula. As an ACS grade Reagent, Spectrum Chemical manufactured Ammonium Hydroxide is used as the quality standard against which other substances are graded and has met the toughest regulatory standards for quality and purity.Specifications
1336-21-6 | |
H5NO | |
UN2672 | |
VHUUQVKOLVNVRT-UHFFFAOYSA-N | |
amine hydrate | |
28 to 30% | |
Poly-Coated Glass Bottle |
100% | |
MFCD00066650 | |
6 x 2.5 L | |
N.O | |
35.05 | |
ACS | |
0.002% |
Chemical Identifiers
1336-21-6 | |
35.05 | |
VHUUQVKOLVNVRT-UHFFFAOYSA-N | |
N.O |
H5NO | |
MFCD00066650 | |
amine hydrate |