missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Hydroxide, Reagent, ACS, 28-30%, Spectrum™ Chemical

A1195, 1336-21-6, NH4OH

Supplier:  Spectrum Chemical Mfg Cor A1195200LTBL

 View more versions of this product

Catalog No. 18600616


Only null left
Explore available promotions

Description

Description

Spectrum™ Chemical Ammonium Hydroxide, Reagent, ACS is a solution of amonia in water. Aqueous amonia, as it′s otherwise known is used as a complexant and base in qualitative inorganic analysis. It becomes a deep blue color with copper solutions. Also used to dissolve silver oxide residues, such as the ones formed from Tollens′ reagent. It is also commmonly used in numerous household cleaning agents, such as window cleaning formula. As an ACS grade Reagent, Spectrum Chemical manufactured Ammonium Hydroxide is used as the quality standard against which other substances are graded and has met the toughest regulatory standards for quality and purity.
Specifications

Specifications

1336-21-6
H5NO
UN2672
VHUUQVKOLVNVRT-UHFFFAOYSA-N
amine hydrate
28 to 30%
Drum
100%
MFCD00066650
200 L
N.O
35.05
ACS
0.002%

Chemical Identifiers

1336-21-6
35.05
VHUUQVKOLVNVRT-UHFFFAOYSA-N
N.O
H5NO
MFCD00066650
amine hydrate
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions