missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium Hydroxide, 2% v/v, Reagents
Ammonium Hydroxide, 2% v/v, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.
$99.47 - $298.33
Chemical Identifiers
CAS | 7732-18-5 |
---|---|
Molecular Formula | H2O |
Molecular Weight (g/mol) | 18.02 |
InChI Key | XLYOFNOQVPJJNP-UHFFFAOYSA-N |
Synonym | Aqua Ammonia |
IUPAC Name | water |
SMILES | O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
01-336-072
|
Labchem Inc
CA0117001A |
1 L |
Each for $99.47
|
|
|||||
01-336-071
|
Labchem Inc
CA0117004A |
4 L |
Each for $298.33
|
|
|||||
Chemical Identifiers
7732-18-5 | |
18.02 | |
Aqua Ammonia | |
O |
H2O | |
XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
water |
Specifications
11.4 | |
89°C | |
H2O | |
1 L | |
O | |
18.02 | |
Reagent | |
0.998 | |
Ammonium Hydroxide, 2% v/v |
Colorless | |
7732-18-5,1336-21-6 | |
Aqua Ammonia | |
XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
water | |
2% | |
Poly Bottle | |
Liquid |