missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium Hydroxide, 10% v/v, Reagents
Ammonium Hydroxide, 10% v/v, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.
Supplier: Labchem Inc CA0115004A
Specifications
>11.5 | |
Colorless | |
7732-18-5,1336-21-6 | |
Aqua Ammonia | |
XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
water | |
10% | |
Poly Bottle | |
Liquid |
Ammonium Hydroxide, 10% v/v | |
69°C | |
H2O | |
4 L | |
O | |
18.02 | |
Reagent | |
0.9892 |
Chemical Identifiers
7732-18-5 | |
18.02 | |
Aqua Ammonia | |
O |
H2O | |
XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
water |