missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium Chloride, TraceSELECT™, Honeywell Fluka™
TraceSELECT™, for trace analysis,≥99.9995% (metals basis)
$672.18 - $672.18
Chemical Identifiers
CAS | 12125-02-9 |
---|---|
Molecular Formula | ClH4N |
Molecular Weight (g/mol) | 53.49 |
MDL Number | MFCD00011420 |
InChI Key | NLXLAEXVIDQMFP-UHFFFAOYSA-N |
Synonym | ammonium chloride, salmiac, sal ammoniac, ammonium muriate, ammoniumchlorid, ammoneric, darammon, amchlor, ammonium chloride nh4 cl, ammoniumklorid |
PubChem CID | 25517 |
ChEBI | CHEBI:31206 |
IUPAC Name | azanium;chloride |
Description
TraceSELECT™ Ultra and TraceSELECT
- ultra-pure acids, bases, and salts for smelting and wet digestion in environmental, water, and food analysis
- sample preparation for trace analysis requires reagents of the highest purity; our TraceSELECT Ultra products for ultra-trace analysis at ppb and even ppt levels are produced by sub-boiling distillation
- the acids, bases, and salts in the TraceSELECT series have been developed for sample preparation and analysis in the ppb (μg/kg) trace range
- packaged in high-quality containers appropriate for each product
- the reagents are produced and bottled under clean-room conditions
Chemical Identifiers
12125-02-9 | |
53.49 | |
NLXLAEXVIDQMFP-UHFFFAOYSA-N | |
25517 | |
azanium;chloride |
ClH4N | |
MFCD00011420 | |
ammonium chloride, salmiac, sal ammoniac, ammonium muriate, ammoniumchlorid, ammoneric, darammon, amchlor, ammonium chloride nh4 cl, ammoniumklorid | |
CHEBI:31206 |
Specifications
12125-02-9 | |
ClH4N | |
MFCD00011420 | |
ammonium chloride, salmiac, sal ammoniac, ammonium muriate, ammoniumchlorid, ammoneric, darammon, amchlor, ammonium chloride nh4 cl, ammoniumklorid | |
[NH4+].[Cl-] | |
53.49 | |
CHEBI:31206 | |
≥99.99% (metals basis) | |
1mmHg (160.4°C) | |
Ammonium Chloride |
100 g | |
NH4Cl | |
4371014 | |
NLXLAEXVIDQMFP-UHFFFAOYSA-N | |
azanium;chloride | |
25517 | |
53.49g/mol | |
TraceSELECT™ | |
Vapor Density: 1.9 (vs air) |
Safety and Handling
P305 + P351 + P338
H302-H319
EINECSNumber : 235-186-4
RTECSNumber : BP4550000