missing translation for 'onlineSavingsMsg'
Learn More

Ammonium chloride, Puriss., meets analytical spec. of BP, FCC, Ph. Eur., USP, 99.5-100.5% (calc. to the dried substance), Honeywell Fluka™

Puriss., meets analytical specification of BP, FCC, Ph. Eur., USP, 99.5-100.5% (calc. to the dried substance)

$374.03 - $1912.25

Chemical Identifiers

CAS 12125-02-9
Molecular Formula ClH4N
Molecular Weight (g/mol) 53.49
MDL Number MFCD00011420
InChI Key NLXLAEXVIDQMFP-UHFFFAOYSA-N
Synonym ammonium chloride, salmiac, sal ammoniac, ammonium muriate, ammoniumchlorid, ammoneric, darammon, amchlor, ammonium chloride nh4 cl, ammoniumklorid
PubChem CID 25517
ChEBI CHEBI:31206
IUPAC Name azanium;chloride
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
6001900
View Documents
Honeywell Chemicals
112091KG
1 kg
Each for $374.03
Only null left
 
6001901
View Documents
Honeywell Chemicals
1120925KG
2.5 kg
Each for $531.13
Only null left
 
60044690
View Documents
Honeywell Chemicals
112096X1KG
6 x 1 kg
Case of 6 Each for $1,912.25
Only null left
 
Description

Description

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications

Chemical Identifiers

12125-02-9
53.49
NLXLAEXVIDQMFP-UHFFFAOYSA-N
25517
azanium;chloride
ClH4N
MFCD00011420
ammonium chloride, salmiac, sal ammoniac, ammonium muriate, ammoniumchlorid, ammoneric, darammon, amchlor, ammonium chloride nh4 cl, ammoniumklorid
CHEBI:31206

Specifications

12125-02-9
ClH4N
MFCD00011420
ammonium chloride, salmiac, sal ammoniac, ammonium muriate, ammoniumchlorid, ammoneric, darammon, amchlor, ammonium chloride nh4 cl, ammoniumklorid
N.Cl
53.49
CHEBI:31206
99.5 to 100.5% (calc. to the dried substance)
≤0.1% (as SO4)
Vapor Density: 1.9 (vs air)
1 kg
NH4Cl
4371014
NLXLAEXVIDQMFP-UHFFFAOYSA-N
azanium;chloride
25517
53.49g/mol
puriss.
1mmHg (160.4°C)
Ammonium Chloride, Meets analytical specification of Ph. Eur.BP, USP, FCC
Videos
Safety and Handling

Safety and Handling

P305 + P351 + P338

H302-H319

EINECSNumber : 235-186-4

RTECSNumber : BP4550000

SDS
missing translation for 'documents'

missing translation for 'documents'