missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium Bisulfate, Crystal, Reagent, 98%, Spectrum™ Chemical
A1140, 7803-63-6, NH4HSO4
$1001.47 - $2099.63
Chemical Identifiers
CAS | 7803-63-6 |
---|---|
Molecular Formula | H5NO4S |
Molecular Weight (g/mol) | 115.10 |
InChI Key | BIGPRXCJEDHCLP-UHFFFAOYSA-N |
IUPAC Name | ammonium hydrogen sulfate |
SMILES | [NH4+].OS([O-])(=O)=O |
Description
Spectrum™ Chemical Ammonium Bisulfate, Crystal, Reagent , also known as ammonium hydrogen sulfate, can be used as an alternative to sulfuric acid. The Reagent grade denotes that this chemical is the highest quality commercially available and that the American Chemical Society has not officially set any specifications for this material. Spectrum Chemical manufactured Reagent grade products meet the toughest regulatory standards for quality and purity.Chemical Identifiers
7803-63-6 | |
115.10 | |
ammonium hydrogen sulfate |
H5NO4S | |
BIGPRXCJEDHCLP-UHFFFAOYSA-N | |
[NH4+].OS([O-])(=O)=O |
Specifications
7803-63-6 | |
100 g | |
UN2506 | |
[NH4+].OS([O-])(=O)=O | |
115.10 | |
Reagent |
100% | |
H5NO4S | |
BIGPRXCJEDHCLP-UHFFFAOYSA-N | |
ammonium hydrogen sulfate | |
98% | |
Amber Glass Bottle |