missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Acetate, For HPLC, Honeywell Fluka™

for HPLC,≥99.0%

$250.35 - $986.50

Chemical Identifiers

CAS 631-61-8
Molecular Formula C2H7NO2
Molecular Weight (g/mol) 77.083
MDL Number MFCD00013066
InChI Key USFZMSVCRYTOJT-UHFFFAOYSA-N
Synonym ammonium acetate, acetic acid, ammonium salt, azanium acetate, ammoniumacetate, acetic acid ammonium salt, ammonium ethanoate, unii-rre756s6q2, aconh4, ch3coonh4, ch3co2nh4
PubChem CID 517165
ChEBI CHEBI:62947
IUPAC Name azanium;acetate
SMILES CC(=O)[O-].[NH4+]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
6002028
View Documents
Honeywell-Fluka
1783650G
50 g N/A
Only null left
 
6002027
View Documents
Honeywell Chemicals
17836250G
250 g N/A
Only null left
 
Description

Description

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications

Chemical Identifiers

631-61-8
77.083
USFZMSVCRYTOJT-UHFFFAOYSA-N
517165
azanium;acetate
C2H7NO2
MFCD00013066
ammonium acetate, acetic acid, ammonium salt, azanium acetate, ammoniumacetate, acetic acid ammonium salt, ammonium ethanoate, unii-rre756s6q2, aconh4, ch3coonh4, ch3co2nh4
CHEBI:62947
CC(=O)[O-].[NH4+]

Specifications

631-61-8
C2H7NO2
MFCD00013066
ammonium acetate, acetic acid, ammonium salt, azanium acetate, ammoniumacetate, acetic acid ammonium salt, ammonium ethanoate, unii-rre756s6q2, aconh4, ch3coonh4, ch3co2nh4
USFZMSVCRYTOJT-UHFFFAOYSA-N
azanium;acetate
517165
77.08g/mol
Ammonium Acetate
50 g
CH3CO2NH4
4186741
Soluble in water 0.1g/mL at 25°C
CC(=O)[O-].[NH4+]
77.083
CHEBI:62947
≥99%
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 211-162-9

RTECSNumber : AF3675000

SDS
missing translation for 'documents'

missing translation for 'documents'