missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium Acetate Buffer, For Iron, Certified, LabChem™
$81.73 - $148.80
Specifications
Chemical Name or Material | Ammonium Acetate Buffer |
---|---|
Name Note | For Iron |
CAS | 7732-18-5,64-19-7,631-61-8 |
CAS Max % | 70.02,20.43,9.55 |
Color | Colorless |
Specifications
Ammonium Acetate Buffer | |
7732-18-5,64-19-7,631-61-8 | |
Colorless | |
Liquid | |
CH3COONH4 | |
Poly Bottle | |
77.08 |
For Iron | |
70.02,20.43,9.55 | |
Gaseous Ammonia | |
Passes Test | |
CH3COONH4 | |
Soluble in water | |
Certified |