missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Ammonia (IIAR) Pipe Markers, Legend: TSS/LIQ/Ammonia/HIGH

For use in labeling ammonia piping and its contents. Brady Ammonia (IIAR) Pipe Markers help create a safer, more efficient work environment.

$26.71 - $26.71

Specifications

Color Black, White, Orange, Red, Yellow
Display Text IIAR Abbreviation: TSS; Physical State: LIQ; Pressure Level: HIGH; Legend: Ammonia
For Use With (Application) Pipe and valve marking
View More Specs

Products 1
Catalog Number Mfr. No. Description Material Diameter (English) Price Quantity  
Catalog Number Mfr. No. Description Material Diameter (English) Price Quantity  
19-110-491
View Documents
Brady™
57999
Style II PVF Over-laminated Polyester (B-689) 2 to 8 in.
Each for $26.71
Only null left
 
Description

Description

  • Meets IIAR Standard and is recommended for use in labeling ammonia piping and its contents
  • Made of B-689 high performance wrap-around PVF over-laminated polyester and B-681/B-883 polyester over-laminate on fiberglass plastic carrier
Specifications

Specifications

Black, White, Orange, Red, Yellow
IIAR Abbreviation: TSS; Physical State: LIQ; Pressure Level: HIGH; Legend: Ammonia
Pipe and valve marking
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'