missing translation for 'onlineSavingsMsg'
Learn More

Amiodarone Hydrochloride 98.0+%, TCI America™

$37.29 - $74.56

Chemical Identifiers

CAS 19774-82-4
Molecular Formula C25H30ClI2NO3
Molecular Weight (g/mol) 681.778
MDL Number MFCD00069204
InChI Key ITPDYQOUSLNIHG-UHFFFAOYSA-N
Synonym amiodarone hydrochloride, amiodarone hcl, amiodar, nexterone, pacerone, amiodaronum hydrochloride, ritmocardyl, rythmarone, angoron
PubChem CID 441325
IUPAC Name (2-butyl-1-benzofuran-3-yl)-[4-[2-(diethylamino)ethoxy]-3,5-diiodophenyl]methanone;hydrochloride
SMILES CCCCC1=C(C2=CC=CC=C2O1)C(=O)C3=CC(=C(C(=C3)I)OCCN(CC)CC)I.Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
A25301G
View Documents
TCI America
A25301G
1 g
Each for $37.29
Only null left
 
A25305G
View Documents
TCI America
A25305G
5 g
Each for $74.56
Only null left
 
Description

Description

Specifications

Chemical Identifiers

19774-82-4
681.778
ITPDYQOUSLNIHG-UHFFFAOYSA-N
441325
CCCCC1=C(C2=CC=CC=C2O1)C(=O)C3=CC(=C(C(=C3)I)OCCN(CC)CC)I.Cl
C25H30ClI2NO3
MFCD00069204
amiodarone hydrochloride, amiodarone hcl, amiodar, nexterone, pacerone, amiodaronum hydrochloride, ritmocardyl, rythmarone, angoron
(2-butyl-1-benzofuran-3-yl)-[4-[2-(diethylamino)ethoxy]-3,5-diiodophenyl]methanone;hydrochloride

Specifications

19774-82-4
White
MFCD00069204
amiodarone hydrochloride, amiodarone hcl, amiodar, nexterone, pacerone, amiodaronum hydrochloride, ritmocardyl, rythmarone, angoron
CCCCC1=C(C2=CC=CC=C2O1)C(=O)C3=CC(=C(C(=C3)I)OCCN(CC)CC)I.Cl
681.778
681.78
Crystalline Powder
156°C
C25H30ClI2NO3
1 g
ITPDYQOUSLNIHG-UHFFFAOYSA-N
(2-butyl-1-benzofuran-3-yl)-[4-[2-(diethylamino)ethoxy]-3,5-diiodophenyl]methanone;hydrochloride
441325
≥98.0% (HPLC,T)
Amiodarone Hydrochloride
Videos
Safety and Handling

Safety and Handling

RTECSNumber : OB1361000

TSCA : No

Recommended Storage : Refrigerator

SDS
missing translation for 'documents'

missing translation for 'documents'