missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ε-Amino-N-caproic Acid, 99.8%, MP Biomedicals™
$82.44 - $82.44
Chemical Identifiers
CAS | 60-32-2 |
---|---|
Molecular Formula | C6H13NO2 |
Molecular Weight (g/mol) | 131.175 |
InChI Key | SLXKOJJOQWFEFD-UHFFFAOYSA-N |
Synonym | 6-aminocaproic acid, aminocaproic acid, amicar, epsikapron, capramol, caprocid, epsamon, eaca, acepramin, caprolisin |
PubChem CID | 564 |
ChEBI | CHEBI:16586 |
IUPAC Name | 6-aminohexanoic acid |
SMILES | C(CCC(=O)O)CCN |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
ICN10164525
|
MP Biomedicals Inc
0210164525 |
25 g |
Each for $82.44
|
|
|||||
Chemical Identifiers
60-32-2 | |
131.175 | |
6-aminocaproic acid, aminocaproic acid, amicar, epsikapron, capramol, caprocid, epsamon, eaca, acepramin, caprolisin | |
CHEBI:16586 | |
C(CCC(=O)O)CCN |
C6H13NO2 | |
SLXKOJJOQWFEFD-UHFFFAOYSA-N | |
564 | |
6-aminohexanoic acid |
Specifications
60-32-2 | |
1.48g/mL | |
C6H13NO2 | |
6-aminocaproic acid, aminocaproic acid, amicar, epsikapron, capramol, caprocid, epsamon, eaca, acepramin, caprolisin | |
C(CCC(=O)O)CCN | |
131.175 | |
CHEBI:16586 | |
Crystalline Powder |
White | |
99.8% | |
25 g | |
SLXKOJJOQWFEFD-UHFFFAOYSA-N | |
6-aminohexanoic acid | |
564 | |
131.2 | |
ε-Amino-N-caproic Acid |