missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aluminum Chloride, Hexahydrate, USP, 95-102%, Spectrum™ Chemical
AL225, 7784-13-6, AlCl3·6H2O
Supplier: Spectrum Chemical Mfg Cor AL22525KG
Description
Spectrum™ Chemical Aluminum Chloride, Hexahydrate, USP is mainly used in topical formulations to treat hyperhidrosis (excessive sweating). All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.Specifications
7784-13-6 | |
2.5 kg | |
MFCD00149134 | |
O.O.O.O.O.O.[Al+3].[Cl-].[Cl-].[Cl-] | |
241.42 | |
USP |
100% | |
AlCl3H12O6 | |
JGDITNMASUZKPW-UHFFFAOYSA-K | |
aluminium(3+) hexahydrate trichloride | |
95 to 102% | |
Poly Pail |
Chemical Identifiers
7784-13-6 | |
241.42 | |
JGDITNMASUZKPW-UHFFFAOYSA-K | |
O.O.O.O.O.O.[Al+3].[Cl-].[Cl-].[Cl-] |
AlCl3H12O6 | |
MFCD00149134 | |
aluminium(3+) hexahydrate trichloride |