missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alphazurine A, MP Biomedicals™
Soluble in water (1mg/mL); Store at Room Temperature (15° to 30°C)
$108.45 - $108.45
Chemical Identifiers
CAS | 3486-30-4 |
---|---|
Molecular Formula | C37H35N2NaO6S2 |
Molecular Weight (g/mol) | 690.805 |
MDL Number | MFCD00044440 |
InChI Key | FTUYQIPAPWPHNC-UHFFFAOYSA-M |
Synonym | acid blue 7, alphazurine a, patent blue a, fenazo bluexg, erio glaucine x, hidacid blue a, carmine blue af, hidacid blue af, kiton blue a, patent blue af |
PubChem CID | 62498 |
IUPAC Name | sodium;4-[[4-[benzyl(ethyl)amino]phenyl]-[4-[benzyl(ethyl)azaniumylidene]cyclohexa-2,5-dien-1-ylidene]methyl]benzene-1,3-disulfonate |
SMILES | CCN(CC1=CC=CC=C1)C2=CC=C(C=C2)C(=C3C=CC(=[N+](CC)CC4=CC=CC=C4)C=C3)C5=C(C=C(C=C5)S(=O)(=O)[O-])S(=O)(=O)[O-].[Na+] |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
ICN15473025
|
MP Biomedicals Inc
0215473025 |
25 g |
Each for $108.45
|
|
|||||
Chemical Identifiers
3486-30-4 | |
690.805 | |
FTUYQIPAPWPHNC-UHFFFAOYSA-M | |
62498 | |
CCN(CC1=CC=CC=C1)C2=CC=C(C=C2)C(=C3C=CC(=[N+](CC)CC4=CC=CC=C4)C=C3)C5=C(C=C(C=C5)S(=O)(=O)[O-])S(=O)(=O)[O-].[Na+] |
C37H35N2NaO6S2 | |
MFCD00044440 | |
acid blue 7, alphazurine a, patent blue a, fenazo bluexg, erio glaucine x, hidacid blue a, carmine blue af, hidacid blue af, kiton blue a, patent blue af | |
sodium;4-[[4-[benzyl(ethyl)amino]phenyl]-[4-[benzyl(ethyl)azaniumylidene]cyclohexa-2,5-dien-1-ylidene]methyl]benzene-1,3-disulfonate |
Specifications
3486-30-4 | |
C37H35N2NaO6S2 | |
25 g | |
FTUYQIPAPWPHNC-UHFFFAOYSA-M | |
sodium;4-[[4-[benzyl(ethyl)amino]phenyl]-[4-[benzyl(ethyl)azaniumylidene]cyclohexa-2,5-dien-1-ylidene]methyl]benzene-1,3-disulfonate | |
62498 | |
Powder |
Purple | |
MFCD00044440 | |
acid blue 7, alphazurine a, patent blue a, fenazo bluexg, erio glaucine x, hidacid blue a, carmine blue af, hidacid blue af, kiton blue a, patent blue af | |
CCN(CC1=CC=CC=C1)C2=CC=C(C=C2)C(=C3C=CC(=[N+](CC)CC4=CC=CC=C4)C=C3)C5=C(C=C(C=C5)S(=O)(=O)[O-])S(=O)(=O)[O-].[Na+] | |
690.805 | |
690.8 | |
Alphazurine A |