missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ansell AlphaTec™ 02-100 Chemical Resistant, 5 Layer Laminated Gloves
Laminated gloves provide extreme resistance against a wide range of chemicals, including biological hazards.
$119.63 - $119.63
Specifications
Certifications/Compliance (Europe) | CE 0493, EN420:2003 + A1:2009; Category lll |
---|---|
Color | White |
Latex Free | Yes |
Material | Laminated Film |
Style | AlphaTec™ 02-100 |
Catalog Number | Mfr. No. | Size | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Size | Price | Quantity | |||||
19-010-058
|
Ansell
902209 |
6 |
Pack of 12 Each for $119.63
Case of 6 Pack for $717.78
|
|
|||||
19-010-059
|
Ansell
0210011070 |
7 |
Pack of 12 Each for $119.63
Case of 6 Pack for $717.78
|
|
|||||
19-010-060
|
Ansell
0210011080 |
8 |
Pack of 12 Each for $119.63
Case of 6 Pack for $717.78
|
|
|||||
19-010-061
|
Ansell
0210011090 |
9 |
Pack of 12 Each for $119.63
Case of 6 Pack for $717.78
|
|
|||||
19-010-062
|
Ansell
988828 |
10 |
Pack of 12 Each for $119.63
Case of 6 Pack for $717.78
|
|
|||||
19-010-063
|
Ansell
0210011110 |
11 |
Pack of 12 Each for $119.63
Case of 6 Pack for $717.78
|
|
|||||
Description
- Provide flexibility and tactility
- Made from an exceptionally thin chemical-resistant, five-layer laminated film
- Integral, nonwoven liner helps absorb and dissipate perspiration for a cooler, more comfortable fit
- Can be worn as liners under heavier nitrile, neoprene, or PVC gloves
- Hand specific
- Broad spectrum chemical resistance
- Wing-thumb for added wearer comfort
Specifications
CE 0493, EN420:2003 + A1:2009; Category lll | |
Yes | |
AlphaTec™ 02-100 | |
0.06 mm | |
No | |
ABCEFGHIMS | |
Gauntlet | |
Unlined | |
15 in. |
White | |
Laminated Film | |
2.5 mil | |
EN 388:0000 | |
Type A | |
Bacteria, Fungi WITH Virus Protection | |
Smooth | |
AlphaTec™ 02-100 | |
38 cm |