missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AlphaProTech Critical Cover™ GenPro™ Frocks
Polypropylene frocks offers protection for non-hazardous environments. Ideal for facility visitors.
$173.94 - $231.09
Specifications
Product Type | Controlled Environments Frock |
---|---|
Material | Composite Fabric |
Color | White |
Sterility | Non-sterile |
Closure Type | Snaps |
Catalog Number | Mfr. No. | Size | Length (Metric) Sleeve | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Size | Length (Metric) Sleeve | Price | Quantity | ||||
19-130-1516B
|
AlphaProTech
FK121222 |
Medium | 87.63 cm |
Case of 30 Each for $199.08
|
|
||||
19-130-1516C
|
AlphaProTech
FK121223 |
Large | 90.17 cm |
Case of 30 Each for $173.94
|
|
||||
19-130-1516D
|
AlphaProTech
FK121224 |
X-Large | 92.71 cm |
Case of 30 Each for $231.09
|
|
||||
19-130-1516F
|
AlphaProTech
FK121226 |
3X-Large | 101.6 cm |
Case of 30 Each for $191.80
|
|
||||
19-130-1516E
|
AlphaProTech
FK121225 |
2X-Large | 95.25 cm |
Case of 30 Each for $228.15
|
|
||||
Description
- Polypropylene frock ideal for personnel or facility visitors
- Offer comfort and breathability
- Extra long design provides additional coverage
- No pockets to help control contaminants from entering or leaving sensitive areas
- Elastic wrist
- Snap mandarin collar
- Five-snap closure
Specifications
Controlled Environments Frock | |
White | |
Snaps | |
Serged | |
Raglan Sleeve |
Composite Fabric | |
Non-sterile | |
Elastic | |
Snap Mandarin |