missing translation for 'onlineSavingsMsg'
Learn More
Learn More
alpha-Sexithiophene (purified by sublimation), TCI America™
$51.02 - $266.46
Chemical Identifiers
CAS | 88493-55-4 |
---|---|
Molecular Formula | C24H14S6 |
Molecular Weight (g/mol) | 494.736 |
InChI Key | KUJYDIFFRDAYDH-UHFFFAOYSA-N |
Synonym | 6T |
PubChem CID | 11340899 |
IUPAC Name | 2-thiophen-2-yl-5-[5-[5-(5-thiophen-2-ylthiophen-2-yl)thiophen-2-yl]thiophen-2-yl]thiophene |
SMILES | C1=CSC(=C1)C2=CC=C(S2)C3=CC=C(S3)C4=CC=C(S4)C5=CC=C(S5)C6=CC=CS6 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
S0504100MG
|
TCI America
S0504100MG |
100 mg |
Each for $51.02
|
|
|||||
S05041G
|
TCI America
S05041G |
1 g |
Each for $266.46
|
|
|||||
Chemical Identifiers
88493-55-4 | |
494.736 | |
6T | |
2-thiophen-2-yl-5-[5-[5-(5-thiophen-2-ylthiophen-2-yl)thiophen-2-yl]thiophen-2-yl]thiophene |
C24H14S6 | |
KUJYDIFFRDAYDH-UHFFFAOYSA-N | |
11340899 | |
C1=CSC(=C1)C2=CC=C(S2)C3=CC=C(S3)C4=CC=C(S4)C5=CC=C(S5)C6=CC=CS6 |
Specifications
88493-55-4 | |
C24H14S6 | |
6T | |
C1=CSC(=C1)C2=CC=C(S2)C3=CC=C(S3)C4=CC=C(S4)C5=CC=C(S5)C6=CC=CS6 | |
494.736 | |
494.74 | |
alpha-Sexithiophene (purified by sublimation) |
304°C | |
100 mg | |
KUJYDIFFRDAYDH-UHFFFAOYSA-N | |
2-thiophen-2-yl-5-[5-[5-(5-thiophen-2-ylthiophen-2-yl)thiophen-2-yl]thiophen-2-yl]thiophene | |
11340899 | |
Crystalline Powder |
Safety and Handling
TSCA : No